Ethyl 6-(1-Pyrrolidyl)pyridine-2-carboxylate
Catalog # APOSOR470045-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:220.268 g/mol
Storage Temperature:Ambient
CAS Number:1166756-92-8
Formula:C₁₂H₁₆N₂O₂
MDL Number:MFCD12198900