
1-tert-Butyl-5H-pyrazolo[3,4-d]pyrimidin-4-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1094377-59-9
:
Formula:C₉H₁₂N₄O
MDL Number:MFCD11207433
:192.218 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together