Order Entry
Italy
ContactUsLinkComponent
1,4-Dibutoxynaphthalene-2,3-dicarbonitrile 99%
Irritant
1,4-Dibutoxynaphthalene-2,3-dicarbonitrile 99%
Codice catalogo APOSOR46229-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,4-Dibutoxynaphthalene-2,3-dicarbonitrile 99%
Codice catalogo APOSOR46229-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :322.401 g/mol
Melting Pt:71…73 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:116453-89-5
Formula:C₂₀H₂₂N₂O₂
MDL Number:MFCD00191997