
6-Formylpyridine-3-boronic acid, pinacol ester
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1073354-14-9
:
Formula:C₁₂H₁₆BNO₃
MDL Number:MFCD06659508
:233.071 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together