
Ethyl 3-azabicyclo[3.1.0]hexane-6-carboxylate hydrochloride 95+%
Catalog # APOSOR45142-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:191.655 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1211510-15-4
Formula:C₈H₁₃NO₂·ClH
MDL Number:MFCD11111227