a-(4-Chlorophenyl)-4-Boc-1-piperazineacetic acid
Catalog # APOSOR451239-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:354.829 g/mol
Storage Temperature:Ambient
CAS Number:885273-01-8
Formula:C₁₇H₂₃ClN₂O₄
MDL Number:MFCD06659337