(R)-4-Boc-2-(aminomethyl)morpholine
Catalog # APOSOR451233-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:216.277 g/mol
Storage Temperature:Ambient
CAS Number:1174913-80-4
Formula:C₁₀H₂₀N₂O₃
MDL Number:MFCD13189480