(S)-2-(1H-Imidazol-1-ylmethyl)-morpholine trihydrochloride
Catalog # APOSOR451232-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:276.591 g/mol
Storage Temperature:Ambient
CAS Number:278789-01-8
Formula:C₈H₁₃N₃O·3ClH
MDL Number:MFCD28975220