Order Entry
Italy
ContactUsLinkComponent
1,4-Dioxaspiro[4.5]decane-8-carboxaldehyde
1,4-Dioxaspiro[4.5]decane-8-carboxaldehyde
Catalog # APOSOR451152-1G
Supplier:  Apollo Scientific
CAS Number:  
1,4-Dioxaspiro[4.5]decane-8-carboxaldehyde
Catalog # APOSOR451152-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :170.206 g/mol
Storage Temperature:Ambient
CAS Number:93245-98-8
Formula:C₉H₁₄O₃
MDL Number:MFCD11847774