Order Entry
Italy
ContactUsLinkComponent
(R)-4-Boc-3-(aminomethyl)-morpholine
(R)-4-Boc-3-(aminomethyl)-morpholine
Catalog # APOSOR451064-1G
Supplier:  Apollo Scientific
CAS Number:  
(R)-4-Boc-3-(aminomethyl)-morpholine
Catalog # APOSOR451064-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :216.277 g/mol
Storage Temperature:Ambient
CAS Number:1187929-33-4
Formula:C₁₀H₂₀N₂O₃
MDL Number:MFCD06739119