Order Entry
Italy
ContactUsLinkComponent
Methyl 5,7-dichloro[1,2,4]triazolo[1,5-a]pyrimidine-2-carboxylate
Methyl 5,7-dichloro[1,2,4]triazolo[1,5-a]pyrimidine-2-carboxylate
Catalog # APOSOR450128-250MG
Supplier:  Apollo Scientific
CAS Number:  
Methyl 5,7-dichloro[1,2,4]triazolo[1,5-a]pyrimidine-2-carboxylate
Catalog # APOSOR450128-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :247.038 g/mol
Storage Temperature:Refrigerator
Formula:C₇H₄Cl₂N₄O₂
MDL Number:MFCD32264092