Order Entry
Italy
ContactUsLinkComponent
5,7-Dichloro[1,2,4]triazolo[1,5-a]pyrimidin-2-ol
5,7-Dichloro[1,2,4]triazolo[1,5-a]pyrimidin-2-ol
Catalog # APOSOR450126-250MG
Supplier:  Apollo Scientific
CAS Number:  
5,7-Dichloro[1,2,4]triazolo[1,5-a]pyrimidin-2-ol
Catalog # APOSOR450126-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :205.002 g/mol
Storage Temperature:Refrigerator
Formula:C₅H₂Cl₂N₄O
MDL Number:MFCD32264090