Order Entry
Italy
ContactUsLinkComponent
5,7-Dichloro-2-nitro[1,2,4]triazolo[1,5-a]pyrimidine
5,7-Dichloro-2-nitro[1,2,4]triazolo[1,5-a]pyrimidine
Catalog # APOSOR450124-250MG
Supplier:  Apollo Scientific
CAS Number:  
5,7-Dichloro-2-nitro[1,2,4]triazolo[1,5-a]pyrimidine
Catalog # APOSOR450124-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :234 g/mol
Storage Temperature:Refrigerator
Formula:C₅HCl₂N₅O₂
MDL Number:MFCD32264089