5,7-Dichloro-2-(methylthio)-[1,2,4]triazolo[1,5-a]pyrimidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1374306-27-0
:
Formula:C₆H₄Cl₂N₄S
MDL Number:MFCD32264083
:235.094 g/mol
Storage Temperature:Refrigerator
Frequently Bought Together