Order Entry
Italy
ContactUsLinkComponent
4-[5-(Ethoxycarbonyl)-4-methyl-1,3-thiazol-2-yl)piperidine, N-BOC protected 90%
4-[5-(Ethoxycarbonyl)-4-methyl-1,3-thiazol-2-yl)piperidine, N-BOC protected 90%
Catalog # APOSOR4426-1G
Supplier:  Apollo Scientific
CAS Number:  
4-[5-(Ethoxycarbonyl)-4-methyl-1,3-thiazol-2-yl)piperidine, N-BOC protected 90%
Catalog # APOSOR4426-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:46.6…47.1 °C
 :354.464 g/mol
Storage Temperature:Ambient
CAS Number:852180-50-8
Formula:C₁₇H₂₆N₂O₄S
MDL Number:MFCD07368516