Order Entry
Italy
ContactUsLinkComponent
Ethyl 1-methyl-1H-indazole-3-carboxylate 97%
Irritant
Ethyl 1-methyl-1H-indazole-3-carboxylate 97%
Catalog # APOSOR4424-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl 1-methyl-1H-indazole-3-carboxylate 97%
Catalog # APOSOR4424-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :204.225 g/mol
Melting Pt:58…63 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:220488-05-1
Formula:C₁₁H₁₂N₂O₂
MDL Number:MFCD07368499