
1-(tert-Butyl) 2-methyl 1H-pyrrole-1,2-dicarboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:294659-30-6
:
Formula:C₁₁H₁₅NO₄
MDL Number:MFCD09842764
:225.241 g/mol
Density:1.0466 g/cm³ (25 °C)
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together