Order Entry
Italy
ContactUsLinkComponent
 
N-(2-Hydroxyethyl)imidazo[1,2-a]pyridine-2-carboxamide
Catalog # APOSOR41008-1G
Supplier:  Apollo Scientific
CAS Number:  
N-(2-Hydroxyethyl)imidazo[1,2-a]pyridine-2-carboxamide
Catalog # APOSOR41008-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :205.213 g/mol
Storage Temperature:Ambient
CAS Number:1157107-85-1
Formula:C₁₀H₁₁N₃O₂
MDL Number:MFCD12179438