
3-[Di(but-1-yl)amino]propan-1-ol
Catalog # APOSOR40320-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:187.322 g/mol
Boiling Pt:94…97 °C (3 mm Hg)
:GHS07
Storage Temperature:Refrigerator
CAS Number:2050-51-3
Formula:C₁₁H₂₅NO
MDL Number:MFCD00152393