Order Entry
Italy
ContactUsLinkComponent
[5-(2-Methyl-1,3-thiazol-4-yl)thien-2-yl]methylamine dihydrochloride
Irritant
[5-(2-Methyl-1,3-thiazol-4-yl)thien-2-yl]methylamine dihydrochloride
Catalog # APOSOR40290-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
[5-(2-Methyl-1,3-thiazol-4-yl)thien-2-yl]methylamine dihydrochloride
Catalog # APOSOR40290-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :283.241 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1030012-47-5
Formula:C₉H₁₀N₂S₂·2ClH
MDL Number:MFCD07704030