Order Entry
Italy
ContactUsLinkComponent
4-Bromo-3-chlorobenzene-1,2-diamine 95%
4-Bromo-3-chlorobenzene-1,2-diamine 95%
Catalog # APOSOR400387-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-3-chlorobenzene-1,2-diamine 95%
Catalog # APOSOR400387-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :221.482 g/mol
Storage Temperature:Ambient
CAS Number:1008361-80-5
Formula:C₆H₆BrClN₂
MDL Number:MFCD22384798