
5-Bromo-3-chlorobenzene-1,2-diamine 97+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16429-44-0
:
Formula:C₆H₆BrClN₂
MDL Number:MFCD12028675
:221.482 g/mol
Melting Pt:58…59 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together