
5-(Phenylethynyl)thiophene-2-carboxaldehyde 97%
Catalog # APOSOR3911-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:212.267 g/mol
Melting Pt:91 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:17257-10-2
Formula:C₁₃H₈OS
MDL Number:MFCD00126374