Order Entry
Italy
ContactUsLinkComponent
2-(2-Methoxyphenyl)-1H-imidazole ≥98%
2-(2-Methoxyphenyl)-1H-imidazole ≥98%
Catalog # APOSOR370070-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(2-Methoxyphenyl)-1H-imidazole ≥98%
Catalog # APOSOR370070-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :174.199 g/mol
Storage Temperature:Ambient
CAS Number:52091-35-7
Formula:C₁₀H₁₀N₂O
MDL Number:MFCD08668846