Order Entry
Italy
ContactUsLinkComponent
4-Bromo-6,7-dimethoxy-indan-1-one ≥98%
4-Bromo-6,7-dimethoxy-indan-1-one ≥98%
Catalog # APOSOR370017-100MG
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-6,7-dimethoxy-indan-1-one ≥98%
Catalog # APOSOR370017-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :271.107 g/mol
Storage Temperature:Ambient
CAS Number:18028-29-0
Formula:C₁₁H₁₁BrO₃
MDL Number:MFCD00088977