Methyl 1-(Hydroxymethyl)cyclopropanecarboxylate ≥98%
Catalog # APOSOR361736-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:130.142 g/mol
Storage Temperature:Ambient
CAS Number:88157-42-0
Formula:C₆H₁₀O₃
MDL Number:MFCD16036399