5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-indazol-3-amine 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:953411-16-0
:
Formula:C₁₃H₁₈BN₃O₂
MDL Number:MFCD16995867
:259.112 g/mol
Storage Temperature:Ambient
Frequently Bought Together