Order Entry
Italy
ContactUsLinkComponent
3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid 97%
3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid 97%
Catalog # APOSOR361481-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid 97%
Catalog # APOSOR361481-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :262.109 g/mol
Storage Temperature:Ambient
CAS Number:269409-74-7
Formula:C₁₄H₁₉BO₄
MDL Number:MFCD08061640