
(S)-6-Methyl-5,6-dihydro-4H-thieno[2,3-b]thiopyran-4-one
Catalog # APOSOR350349-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:184.279 g/mol
Melting Pt:43…45 °C
Density:1.3 g/cm³ (25 °C)
Flash Pt:140 °C
:GHS07
Storage Temperature:Ambient
CAS Number:147086-79-1
Formula:C₈H₈OS₂
MDL Number:MFCD09027305