Order Entry
Italy
ContactUsLinkComponent
2,6-Dibromo-4-chlorophenol
Toxic
2,6-Dibromo-4-chlorophenol
Catalog # APOSOR350143-1G
Supplier:  Apollo Scientific
CAS Number:  
Toxic
2,6-Dibromo-4-chlorophenol
Catalog # APOSOR350143-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :286.348 g/mol
Melting Pt:93 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:5324-13-0
Formula:C₆H₃Br₂ClO
MDL Number:MFCD00029751