Order Entry
Italy
ContactUsLinkComponent
 
(2,5-Dibromophenyl)hydrazine 97%
Catalog # APOSOR350136-1G
Supplier:  Apollo Scientific
CAS Number:  
(2,5-Dibromophenyl)hydrazine 97%
Catalog # APOSOR350136-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :265.933 g/mol
Storage Temperature:Ambient
CAS Number:62672-26-8
Formula:C₆H₆Br₂N₂
MDL Number:MFCD02663135