(5-Ethylsulphanyl-[1,3,4]oxadiazol-2-ylmethyl)-carbamic acid tert-butyl ester
Catalog # APOSOR346349-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:259.325 g/mol
Storage Temperature:Ambient
CAS Number:914216-58-3
Formula:C₁₀H₁₇N₃O₃S
MDL Number:MFCD00436038