Order Entry
Italy
ContactUsLinkComponent
2-Amino-3-chloro-4-methylpyridine ≥97%
2-Amino-3-chloro-4-methylpyridine ≥97%
Catalog # APOSOR345371-250MG
Supplier:  Apollo Scientific
CAS Number:  
2-Amino-3-chloro-4-methylpyridine ≥97%
Catalog # APOSOR345371-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :142.586 g/mol
Storage Temperature:Refrigerator
CAS Number:56960-76-0
Formula:C₆H₇ClN₂
MDL Number:MFCD09839234