
2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:104617-49-4
:
Formula:C₇H₁₁N₃S
MDL Number:MFCD07771983
:169.247 g/mol
Melting Pt:230…231 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together