Order Entry
Italy
ContactUsLinkComponent
 
(R)-3,3'-Bis(anthracenyl-9-yl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Catalog # APOSOR340032-250MG
Supplier:  Apollo Scientific
CAS Number:  
(R)-3,3'-Bis(anthracenyl-9-yl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Catalog # APOSOR340032-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :700.715 g/mol
Melting Pt:260…274 °C
Storage Temperature:Freezer
CAS Number:361342-51-0
Formula:C₄₈H₂₉O₄P
MDL Number:MFCD11045406