Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dimethyl-2-[(morpholin-4-yl)methyl]phenol
3,5-Dimethyl-2-[(morpholin-4-yl)methyl]phenol, tech
3,5-Dimethyl-2-[(morpholin-4-yl)methyl]phenol, tech
APOSOR33434-1G
:
Apollo Scientific
:
3,5-Dimethyl-2-[(morpholin-4-yl)methyl]phenol, tech
APOSOR33434-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
221.295 g/mol
:
Ambient
:
14944-83-3
:
C₁₃H₁₉NO₂
:
MFCD05668993