Methyl 4-(2-methylbenzoyl)-1H-pyrrole-2-carboxylate, tech
Catalog # APOSOR33392-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:243.258 g/mol
Storage Temperature:Ambient
CAS Number:478249-31-9
Formula:C₁₄H₁₃NO₃
MDL Number:MFCD02571494