Order Entry
Italy
ContactUsLinkComponent
 
2-(Benzylamino)-3-chloro-1,4-dihydronaphthalene-1,4-dione, tech
Catalog # APOSOR33389-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(Benzylamino)-3-chloro-1,4-dihydronaphthalene-1,4-dione, tech
Catalog # APOSOR33389-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :297.736 g/mol
Storage Temperature:Ambient
CAS Number:4497-69-2
Formula:C₁₇H₁₂ClNO₂
MDL Number:MFCD00169805