Order Entry
Italy
ContactUsLinkComponent
3-Benzyl-4-hydroxy-1-(prop-2-yn-1-yl)-1,2-dihydropyridin-2-one, tech
3-Benzyl-4-hydroxy-1-(prop-2-yn-1-yl)-1,2-dihydropyridin-2-one, tech
Catalog # APOSOR33384-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Benzyl-4-hydroxy-1-(prop-2-yn-1-yl)-1,2-dihydropyridin-2-one, tech
Catalog # APOSOR33384-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :239.269 g/mol
Storage Temperature:Ambient
CAS Number:478247-72-2
Formula:C₁₅H₁₃NO₂
MDL Number:MFCD01871744