Order Entry
Italy
ContactUsLinkComponent
3-Chloro-N-(4-cyanophenyl)-2,2-dimethylpropanamide, tech
3-Chloro-N-(4-cyanophenyl)-2,2-dimethylpropanamide, tech
Catalog # APOSOR33363-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Chloro-N-(4-cyanophenyl)-2,2-dimethylpropanamide, tech
Catalog # APOSOR33363-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :236.697 g/mol
Storage Temperature:Ambient
CAS Number:478261-84-6
Formula:C₁₂H₁₃ClN₂O
MDL Number:MFCD00793439