Order Entry
Italy
ContactUsLinkComponent
1-[4-Methyl-2-(morpholin-4-yl)quinolin-3-yl]ethan-1-one, tech
1-[4-Methyl-2-(morpholin-4-yl)quinolin-3-yl]ethan-1-one, tech
Catalog # APOSOR33259-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[4-Methyl-2-(morpholin-4-yl)quinolin-3-yl]ethan-1-one, tech
Catalog # APOSOR33259-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :270.326 g/mol
Storage Temperature:Ambient
CAS Number:339106-15-9
Formula:C₁₆H₁₈N₂O₂
MDL Number:MFCD01568465