Order Entry
Italy
ContactUsLinkComponent
5H,6H,7H,8H-Imidazo[1,5-c]pyrimidin-5-one, tech
5H,6H,7H,8H-Imidazo[1,5-c]pyrimidin-5-one, tech
Catalog # APOSOR33250-1G
Supplier:  Apollo Scientific
CAS Number:  
5H,6H,7H,8H-Imidazo[1,5-c]pyrimidin-5-one, tech
Catalog # APOSOR33250-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :137.139 g/mol
Storage Temperature:Ambient
CAS Number:14509-66-1
Formula:C₆H₇N₃O
MDL Number:MFCD01443896