Order Entry
Italy
ContactUsLinkComponent
2-(2,4-Dichlorophenyl)-2-(morpholin-4-yl)acetonitrile, tech
2-(2,4-Dichlorophenyl)-2-(morpholin-4-yl)acetonitrile, tech
Catalog # APOSOR33234-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(2,4-Dichlorophenyl)-2-(morpholin-4-yl)acetonitrile, tech
Catalog # APOSOR33234-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :271.142 g/mol
Storage Temperature:Ambient
CAS Number:135595-04-9
Formula:C₁₂H₁₂Cl₂N₂O
MDL Number:MFCD00172631