Order Entry
Italy
ContactUsLinkComponent
N-(4-Methoxyphenyl)-2-oxo-1-(prop-2-yn-1-yl)-1,2-dihydropyridine-3-carboxamide, tech
N-(4-Methoxyphenyl)-2-oxo-1-(prop-2-yn-1-yl)-1,2-dihydropyridine-3-carboxamide, tech
Catalog # APOSOR33219-1G
Supplier:  Apollo Scientific
CAS Number:  
N-(4-Methoxyphenyl)-2-oxo-1-(prop-2-yn-1-yl)-1,2-dihydropyridine-3-carboxamide, tech
Catalog # APOSOR33219-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :282.294 g/mol
Storage Temperature:Ambient
CAS Number:339028-30-7
Formula:C₁₆H₁₄N₂O₃
MDL Number:MFCD00141338