Order Entry
Italy
ContactUsLinkComponent
Methyl 2-(2,5-dimethyl-1H-pyrrol-1-yl)thiophene-3-carboxylate, tech
Methyl 2-(2,5-dimethyl-1H-pyrrol-1-yl)thiophene-3-carboxylate, tech
Catalog # APOSOR33142-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2-(2,5-dimethyl-1H-pyrrol-1-yl)thiophene-3-carboxylate, tech
Catalog # APOSOR33142-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :235.302 g/mol
Storage Temperature:Ambient
CAS Number:478077-85-9
Formula:C₁₂H₁₃NO₂S
MDL Number:MFCD01312398