Order Entry
Italy
ContactUsLinkComponent
(2Z)-2-(2,4-Dichlorophenyl)-3-hydroxyprop-2-enenitrile, tech
(2Z)-2-(2,4-Dichlorophenyl)-3-hydroxyprop-2-enenitrile, tech
Catalog # APOSOR33075-1G
Supplier:  Apollo Scientific
CAS Number:  
(2Z)-2-(2,4-Dichlorophenyl)-3-hydroxyprop-2-enenitrile, tech
Catalog # APOSOR33075-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :214.048 g/mol
Storage Temperature:Ambient
CAS Number:115716-98-8
Formula:C₉H₅Cl₂NO
MDL Number:MFCD00141627