Order Entry
Italy
ContactUsLinkComponent
2-[3-(2H-1,3-Benzodioxol-5-yl)-1H-pyrazol-1-yl]ethan-1-ol, tech
2-[3-(2H-1,3-Benzodioxol-5-yl)-1H-pyrazol-1-yl]ethan-1-ol, tech
Catalog # APOSOR33031-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[3-(2H-1,3-Benzodioxol-5-yl)-1H-pyrazol-1-yl]ethan-1-ol, tech
Catalog # APOSOR33031-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :232.235 g/mol
Storage Temperature:Ambient
CAS Number:955976-57-5
Formula:C₁₂H₁₂N₂O₃
MDL Number:MFCD03617679