Order Entry
Italy
ContactUsLinkComponent
N'-[(1E)-(Dimethylamino)methylidene]-1H,2H-naphtho[2,1-b]furan-2-carbohydrazide, tech
N'-[(1E)-(Dimethylamino)methylidene]-1H,2H-naphtho[2,1-b]furan-2-carbohydrazide, tech
Catalog # APOSOR33022-1G
Supplier:  Apollo Scientific
CAS Number:  
N'-[(1E)-(Dimethylamino)methylidene]-1H,2H-naphtho[2,1-b]furan-2-carbohydrazide, tech
Catalog # APOSOR33022-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :283.325 g/mol
Storage Temperature:Ambient
CAS Number:478064-24-3
Formula:C₁₆H₁₇N₃O₂
MDL Number:MFCD02187225