Order Entry
Italy
ContactUsLinkComponent
5-(Methoxymethyl)-2-methylpyrazolo[1,5-a]pyrimidin-7-ol, tech
5-(Methoxymethyl)-2-methylpyrazolo[1,5-a]pyrimidin-7-ol, tech
Catalog # APOSOR33018-1G
Supplier:  Apollo Scientific
CAS Number:  
5-(Methoxymethyl)-2-methylpyrazolo[1,5-a]pyrimidin-7-ol, tech
Catalog # APOSOR33018-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :193.203 g/mol
Storage Temperature:Ambient
CAS Number:439111-05-4
Formula:C₉H₁₁N₃O₂
MDL Number:MFCD02661789