Order Entry
Italy
ContactUsLinkComponent
Ethyl 3-hydroxypyrido[1,2-a]indole-10-carboxylate, tech
Ethyl 3-hydroxypyrido[1,2-a]indole-10-carboxylate, tech
Catalog # APOSOR32999-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 3-hydroxypyrido[1,2-a]indole-10-carboxylate, tech
Catalog # APOSOR32999-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :255.269 g/mol
Storage Temperature:Ambient
CAS Number:22991-17-9
Formula:C₁₅H₁₃NO₃
MDL Number:MFCD01443652